Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rqcd1
Entrez Gene ID
58184
Gene Name
rcd1 (required for cell differentiation) homolog 1 (S. pombe)
Gene Aliases
2610007F23Rik, AI593551, Fl10
Chromosome Location
Chr.1: 74506040 - 74530842 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_021383.5 NP_067358.1 - 403 71
GenBank mRNA AK005025.2 - 1 - 2 405 71
BC050898.1 - 1 - 2 204 71
BC051948.1 - - 242 71
BY705231.1 - 1 - 2 403 71

View Full Product Details