Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Isl1
Entrez Gene ID
16392
Gene Name
ISL1 transcription factor, LIM/homeodomain
Gene Aliases
-
Chromosome Location
Chr.13: 116298257 - 116309721 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_021459.4 NP_067434.3 2 - 3 485 99
GenBank mRNA AB104633.1 - 2 - 3 219 99
AJ132765.1 - 2 - 3 351 99
AK090263.1 - 2 - 3 486 99
BC132263.1 - 2 - 3 411 99
BC132609.1 - 2 - 3 411 99
CF725499.1 - 1 - 2 120 99

View Full Product Details