The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009430.2 | NP_033456.1 | 2 - 3 | 234 | 131 |
GenBank mRNA | AK003082.1 | - | 2 - 3 | 223 | 131 |
BB841364.1 | - | 2 - 3 | 235 | 131 | |
BC064796.1 | - | 2 - 3 | 210 | 131 | |
X04574.1 | - | 2 - 3 | 222 | 131 |