Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ttf1
Entrez Gene ID
22130
Gene Name
transcription termination factor, RNA polymerase I
Gene Aliases
AV245725, TTF-1, TTF-I
Chromosome Location
Chr.2: 29060199 - 29087656 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009442.2 NP_033468.2 5 - 6 1826 115
GenBank mRNA AF237703.1 - 4 - 5 1778 115
AK140081.1 - 4 - 5 3044 115
BC059011.1 - 4 - 5 1793 115
X83974.1 - 6 - 7 1734 115

View Full Product Details