The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_023579.4 | NP_076068.1 | 4 - 5 | 528 | 121 |
GenBank mRNA | AF294327.1 | - | 4 - 5 | 479 | 121 |
AK017701.1 | - | 4 - 5 | 487 | 121 | |
AK049422.1 | - | 4 - 5 | 500 | 121 | |
AK053681.1 | - | 4 - 5 | 493 | 121 | |
AK079134.1 | - | 4 - 5 | 498 | 121 | |
AK167303.1 | - | 4 - 5 | 490 | 121 | |
AK169664.1 | - | 4 - 5 | 496 | 121 | |
BC052392.1 | - | 4 - 5 | 471 | 121 | |
BC054814.1 | - | 4 - 5 | 701 | 121 | |
CB524964.1 | - | 4 - 5 | 487 | 121 |