Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ttc7
Entrez Gene ID
225049
Gene Name
tetratricopeptide repeat domain 7
Gene Aliases
1110035E02Rik, 1700007L07Rik, Ttc7a, fsn, hea
Chromosome Location
Chr.17: 87282082 - 87381770 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_028639.3 NP_082915.1 16 - 17 2261 70
GenBank mRNA AK040578.1 - 16 - 17 2264 70
AK040610.1 - 16 - 17 2232 70
AK050263.1 - 16 - 17 2248 70
AK080284.1 - 16 - 17 2240 70
AK153704.1 - 16 - 17 2243 70
AK165244.1 - 9 - 10 923 70
BC023773.1 - 12 - 13 1219 70
BC042512.1 - 16 - 17 1938 70

View Full Product Details