The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001177696.1 | NP_001171167.1 | 13 - 14 | 1501 | 118 |
NM_001177697.1 | NP_001171168.1 | 13 - 14 | 1477 | 118 | |
NM_001177698.1 | NP_001171169.1 | 13 - 14 | 1501 | 118 | |
NM_001177699.1 | NP_001171170.1 | 12 - 13 | 1444 | 118 | |
GenBank mRNA | AK046879.1 | - | 13 - 14 | 1426 | 118 |
AK082707.1 | - | 12 - 13 | 1394 | 118 | |
AK154494.1 | - | 13 - 14 | 1503 | 118 | |
AY274116.1 | - | 12 - 13 | 1371 | 118 | |
BC141125.1 | - | 12 - 13 | 1304 | 118 | |
DQ413175.1 | - | 12 - 13 | 1330 | 118 |