The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_177708.5 | NP_808376.1 | 1 - 2 | 303 | 52 |
GenBank mRNA | AK033286.1 | - | 1 - 2 | 301 | 52 |
AK140456.1 | - | 1 - 2 | 304 | 52 | |
BC030471.1 | - | 1 - 2 | 507 | 52 |