Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cirh1a
Entrez Gene ID
21771
Gene Name
cirrhosis, autosomal recessive 1A (human)
Gene Aliases
Cirhin, Naic, Teg-292, Tex292
Chromosome Location
Chr.8: 106893636 - 106923094 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011574.2 NP_035704.2 12 - 13 1515 71
GenBank mRNA AK144455.1 - 12 - 13 1500 71
AK146263.1 - 12 - 13 1491 71
AK150862.1 - 12 - 13 1491 71
AK168470.1 - 12 - 13 1465 71
AK173318.1 - 11 - 12 1456 71
BC027399.1 - 8 - 9 1014 71
BC068113.1 - 12 - 13 1504 71

View Full Product Details