The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_027290.3 | NP_081566.2 | 6 - 7 | 984 | 66 |
GenBank mRNA | AK030780.1 | - | 6 - 7 | 929 | 66 |
AK041406.1 | - | 6 - 7 | 919 | 66 | |
BC064314.1 | - | 6 - 7 | 1347 | 66 | |
BC094576.1 | - | 6 - 7 | 984 | 66 | |
BC120687.1 | - | 5 - 6 | 893 | 66 | |
BC120689.1 | - | 5 - 6 | 893 | 66 |