Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dtl
Entrez Gene ID
76843
Gene Name
denticleless homolog (Drosophila)
Gene Aliases
2810047L02Rik, 5730564G15Rik, Ramp
Chromosome Location
Chr.1: 191537365 - 191575534 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AB095735.1 - 7 - 8 755 91
AB095736.1 - 7 - 8 755 91
AK054412.1 - 7 - 8 745 91
AK133177.1 - 7 - 8 754 91
AK154594.1 - 8 - 9 795 91
AK161401.1 - 4 - 5 356 91
AK164249.1 - 7 - 8 755 91
AK164999.1 - 7 - 8 754 91
AK166351.1 - 7 - 8 763 91
AK169601.1 - 7 - 8 754 91
BC060208.1 - 7 - 8 714 91

View Full Product Details