Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Kif27
Entrez Gene ID
75050
Gene Name
kinesin family member 27
Gene Aliases
4930517I18Rik
Chromosome Location
Chr.13: 58287516 - 58364745 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_175214.3 NP_780423.2 1 - 2 292 71

View Full Product Details