Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ubash3b
Entrez Gene ID
72828
Gene Name
ubiquitin associated and SH3 domain containing, B
Gene Aliases
2810457I06Rik, BB125008, TULA-2, p70
Chromosome Location
Chr.9: 41010760 - 41161692 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_176860.5 NP_789830.1 2 - 3 250 105
GenBank mRNA AB075602.1 - 2 - 3 245 105
AK013361.1 - 2 - 3 318 105
AK034450.1 - 2 - 3 441 105
AK035764.1 - 2 - 3 251 105
AK133948.1 - 2 - 3 248 105
AK150909.1 - 2 - 3 818 104
AK151721.1 - 2 - 3 818 104
AK151895.1 - 2 - 3 451 105
AK152013.1 - 2 - 3 451 105
AK154576.1 - 2 - 3 472 105
AK172288.1 - - 251 105
BC053436.1 - 2 - 3 222 105

View Full Product Details