The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_176860.5 | NP_789830.1 | 3 - 4 | 432 | 82 |
GenBank mRNA | AB075602.1 | - | 3 - 4 | 427 | 82 |
AK013361.1 | - | 3 - 4 | 500 | 82 | |
AK034450.1 | - | 3 - 4 | 623 | 82 | |
AK035764.1 | - | 3 - 4 | 433 | 82 | |
AK133948.1 | - | 3 - 4 | 430 | 82 | |
AK150909.1 | - | 3 - 4 | 999 | 82 | |
AK151721.1 | - | 3 - 4 | 1000 | 82 | |
AK151895.1 | - | 3 - 4 | 633 | 82 | |
AK152013.1 | - | 3 - 4 | 633 | 82 | |
AK154576.1 | - | 3 - 4 | 654 | 82 | |
AK172288.1 | - | - | 433 | 82 | |
BC053436.1 | - | 3 - 4 | 404 | 82 |