The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_027225.1 | NP_081501.1 | 2 - 3 | 449 | 66 |
NM_177025.5 | NP_795999.2 | 2 - 3 | 449 | 66 | |
GenBank mRNA | AK037010.1 | - | 3 - 4 | 440 | 66 |
AK052533.1 | - | 2 - 3 | 398 | 66 | |
AK144943.1 | - | 3 - 4 | 493 | 66 | |
AK164624.1 | - | 3 - 4 | 379 | 66 | |
AK173076.1 | - | 2 - 3 | 280 | 66 | |
AY308746.1 | - | 2 - 3 | 449 | 66 | |
BC067007.1 | - | 2 - 3 | 446 | 66 |