Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Adamts20
Entrez Gene ID
223838
Gene Name
a disintegrin-like and metallopeptidase (reprolysin type) with thrombospondin type 1 motif, 20
Gene Aliases
Adamst20, bt
Chromosome Location
Chr.15: 94268314 - 94465427 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001164785.1 NP_001158257.1 5 - 6 1200 129
NM_001164786.1 NP_001158258.1 5 - 6 1200 129
NM_177431.4 NP_803180.3 5 - 6 1200 129
GenBank mRNA AJ512753.3 - 5 - 6 969 129
AK090358.1 - 5 - 6 1166 129
AY189815.1 - 5 - 6 1108 129
AY189816.1 - 5 - 6 1108 129
BC121792.1 - 5 - 6 1039 129

View Full Product Details