The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_178600.2 | NP_848715.1 | 2 - 3 | 323 | 68 |
GenBank mRNA | AK002742.1 | - | 2 - 3 | 304 | 68 |
AK003237.1 | - | 2 - 3 | 324 | 68 | |
AK008509.1 | - | 2 - 3 | 297 | 68 | |
AK008578.1 | - | 2 - 3 | 298 | 68 | |
AK013996.1 | - | 2 - 3 | 307 | 68 | |
BC031732.1 | - | 2 - 3 | 305 | 68 | |
CD774913.1 | - | 1 - 2 | 68 |