Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Gad1
- Entrez Gene ID
-
14415
- Gene Name
- glutamate decarboxylase 1
- Gene Aliases
-
EP10, GAD25, GAD44, GAD67, Gad-1
- Chromosome Location
-
Chr.2: 70561763 - 70602014 on Build GRCm38
- Gene Symbol
- Gad1-ps
- Entrez Gene ID
-
382407
- Gene Name
- glutamate decarboxylase 1, pseudogene
- Gene Aliases
-
Gad-1ps
- Chromosome Location
-
Chr.10: 99443838 - 99447029 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK137453.1 |
- |
1 - 1 |
1875 |
66 |
AK137640.1 |
- |
1 - 1 |
1899 |
66 |
View Full Product Details