Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Sf3b4
Entrez Gene ID
107701
Gene Name
splicing factor 3b, subunit 4
Gene Aliases
SF3b49, SF3b50, Sap49
Chromosome Location
Chr.3: 96172550 - 96177564 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK047751.1 - 3 - 3 581 81
AK135543.1 - 3 - 3 589 81
AK156828.1 - 3 - 3 586 81
AK205335.1 - 1 - 1 158 81
BC024418.1 - 3 - 3 624 81
BC026567.1 - 3 - 3 612 81
BC085273.1 - 3 - 3 564 81

View Full Product Details