The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_181817.1 | NP_861538.1 | 1 - 1 | 554 | 93 |
GenBank mRNA | AY288422.1 | - | 1 - 1 | 554 | 93 |
AY633765.1 | - | 1 - 1 | 554 | 93 | |
BC107269.1 | - | 1 - 1 | 93 | ||
BC107270.1 | - | 1 - 1 | 93 |