The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001163701.1 | NP_001157173.1 | 2 - 3 | 360 | 72 |
NM_177294.5 | NP_796268.3 | 3 - 4 | 437 | 72 | |
GenBank mRNA | AK042776.1 | - | 2 - 3 | 338 | 72 |
AK122505.1 | - | 3 - 4 | 404 | 72 | |
AK154007.1 | - | 2 - 3 | 343 | 72 | |
AK161487.1 | - | 3 - 4 | 420 | 72 | |
AK164658.1 | - | 3 - 4 | 420 | 72 | |
AK166232.1 | - | 2 - 3 | 343 | 72 | |
AK167589.1 | - | 2 - 3 | 343 | 72 | |
AK171233.1 | - | 2 - 3 | 316 | 72 | |
BC088899.1 | - | 2 - 3 | 323 | 72 | |
BC117973.1 | - | 2 - 3 | 329 | 72 | |
BC117974.1 | - | 2 - 3 | 329 | 72 |