The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_146243.2 | NP_666355.1 | 3 - 4 | 473 | 94 |
GenBank mRNA | AK032832.1 | - | 3 - 4 | 391 | 94 |
AK033014.1 | - | 3 - 4 | 391 | 94 | |
AK088216.1 | - | 3 - 4 | 385 | 94 | |
AK163217.1 | - | 3 - 4 | 386 | 94 | |
BC027799.1 | - | 3 - 4 | 457 | 94 | |
BC047530.1 | - | 3 - 4 | 372 | 94 |