The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_172253.2 | NP_758457.1 | 3 - 4 | 631 | 110 |
GenBank mRNA | AK008677.1 | - | 3 - 4 | 613 | 110 |
AK010829.1 | - | 3 - 4 | 632 | 110 | |
AK016638.1 | - | 3 - 4 | 628 | 110 | |
AK017016.1 | - | 3 - 4 | 109 | ||
AK017810.1 | - | 3 - 4 | 616 | 110 | |
AK153090.1 | - | - | 621 | 110 | |
AK160682.1 | - | 3 - 4 | 619 | 110 | |
BC082580.1 | - | - | 603 | 110 | |
BC139162.1 | - | 3 - 4 | 619 | 110 | |
BC139163.1 | - | 3 - 4 | 619 | 110 |