The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_145625.3 | NP_663600.2 | 9 - 9 | 1250 | 151 |
GenBank mRNA | AK045250.1 | - | 9 - 9 | 1094 | 151 |
AK077423.1 | - | 9 - 9 | 1101 | 151 | |
AK148012.1 | - | 9 - 9 | 1100 | 151 | |
AK161693.1 | - | 9 - 9 | 1083 | 151 | |
AK166363.1 | - | 9 - 9 | 1102 | 151 | |
AK168350.1 | - | 9 - 9 | 1096 | 151 | |
AK168425.1 | - | 9 - 9 | 1096 | 151 | |
AK170256.1 | - | 9 - 9 | 1101 | 151 | |
AK170354.1 | - | 9 - 9 | 1083 | 151 | |
BC007171.1 | - | 8 - 8 | 1049 | 151 | |
BC036293.1 | - | 9 - 9 | 1086 | 151 | |
BC147746.1 | - | 9 - 9 | 1159 | 151 | |
BC147748.1 | - | 9 - 9 | 1159 | 151 |