The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_178616.3 | NP_848731.2 | 10 - 10 | 1054 | 66 |
GenBank mRNA | AK007547.1 | - | 10 - 10 | 1023 | 66 |
AK012951.1 | - | 10 - 10 | 1063 | 66 | |
AK029365.1 | - | 6 - 6 | 625 | 66 | |
AK051738.1 | - | 10 - 10 | 1044 | 66 | |
AK076002.1 | - | 10 - 10 | 1042 | 66 | |
AK089942.1 | - | 10 - 10 | 1019 | 66 | |
AK136049.1 | - | 10 - 10 | 1017 | 66 | |
AK140773.1 | - | - | 1039 | 66 | |
AK157616.1 | - | 1 - 1 | 66 | ||
AK167559.1 | - | 10 - 10 | 1025 | 66 | |
AK168289.1 | - | 10 - 10 | 1046 | 66 | |
BC030432.1 | - | 7 - 7 | 708 | 66 | |
BC037590.1 | - | 10 - 10 | 1003 | 66 | |
BC055457.1 | - | 10 - 10 | 986 | 66 | |
BC090664.1 | - | 10 - 10 | 1006 | 66 | |
BC090980.1 | - | 10 - 10 | 1021 | 66 | |
BC100595.1 | - | 10 - 10 | 1016 | 66 | |
BC119136.1 | - | 10 - 10 | 1025 | 66 | |
BC119138.1 | - | 10 - 10 | 1025 | 66 |