The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_028622.2 | NP_082898.1 | 2 - 2 | 548 | 105 |
GenBank mRNA | AK003705.1 | - | 2 - 2 | 549 | 105 |