The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001301301.1 | NP_001288230.1 | 4 - 5 | 431 | 80 |
NM_001301303.1 | NP_001288232.1 | 4 - 5 | 418 | 80 | |
NM_001301305.1 | NP_001288234.1 | 4 - 5 | 410 | 80 | |
NM_001301308.1 | NP_001288237.1 | 3 - 4 | 260 | 80 | |
NM_001301309.1 | NP_001288238.1 | 3 - 4 | 241 | 80 | |
NM_007486.5 | NP_031512.1 | 4 - 5 | 450 | 80 | |
GenBank mRNA | AA219822.1 | - | 4 - 5 | 384 | 80 |
AK002516.1 | - | 4 - 5 | 412 | 80 | |
AK169733.1 | - | 4 - 5 | 408 | 80 | |
BB755757.1 | - | 4 - 5 | 400 | 80 | |
BC031763.1 | - | 4 - 5 | 363 | 80 | |
L07918.1 | - | 4 - 5 | 364 | 80 |