Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pcsk4
Entrez Gene ID
18551
Gene Name
proprotein convertase subtilisin/kexin type 4
Gene Aliases
AI647044, L21221, NEC 3, NEC3, PC4, SPC5
Chromosome Location
Chr.10: 80321281 - 80329909 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008793.2 NP_032819.1 7 - 8 870 74
GenBank mRNA D01093.1 - 7 - 8 870 74

View Full Product Details