The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009618.2 | NP_033748.2 | 15 - 16 | 1634 | 123 |
GenBank mRNA | AK016550.1 | - | 15 - 16 | 1634 | 123 |
AK161266.1 | - | 15 - 16 | 1623 | 123 | |
U16242.1 | - | 15 - 16 | 1623 | 123 | |
U22057.1 | - | 14 - 15 | 1574 | 123 | |
U38806.1 | - | 15 - 16 | 1639 | 123 |