The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001253886.1 | NP_001240815.1 | 8 - 9 | 1750 | 94 |
NM_009685.3 | NP_033815.1 | 8 - 9 | 1564 | 94 | |
GenBank mRNA | AF206720.1 | - | 8 - 9 | 1473 | 94 |
AK030748.1 | - | 8 - 9 | 1447 | 94 |