The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001201413.1 | NP_001188342.1 | 16 - 17 | 2685 | 75 |
NM_001201414.1 | NP_001188343.1 | 15 - 16 | 2688 | 75 | |
NM_001201415.1 | NP_001188344.1 | 15 - 16 | 2622 | 75 | |
NM_001201416.1 | NP_001188345.1 | 6 - 7 | 788 | 75 | |
NM_009686.2 | NP_033816.1 | 17 - 18 | 2691 | 75 | |
GenBank mRNA | AF133665.1 | - | 2 - 3 | 263 | 75 |
AK004792.1 | - | 17 - 18 | 2545 | 75 | |
AK153260.1 | - | 15 - 16 | 2654 | 75 | |
AK167649.1 | - | 16 - 17 | 2550 | 75 | |
AK169959.1 | - | 15 - 16 | 2560 | 75 | |
AK172405.1 | - | 6 - 7 | 792 | 75 | |
BC042797.1 | - | 7 - 8 | 830 | 75 | |
BC076587.1 | - | 15 - 16 | 2468 | 75 |