The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001201413.1 | NP_001188342.1 | 9 - 10 | 1818 | 106 |
NM_001201414.1 | NP_001188343.1 | 8 - 9 | 1821 | 106 | |
NM_001201415.1 | NP_001188344.1 | 8 - 9 | 1755 | 106 | |
NM_009686.2 | NP_033816.1 | 9 - 10 | 1818 | 106 | |
GenBank mRNA | AK004792.1 | - | 9 - 10 | 1672 | 106 |
AK153260.1 | - | 8 - 9 | 106 | ||
AK167649.1 | - | 9 - 10 | 1683 | 106 | |
AK169959.1 | - | 8 - 9 | 1696 | 106 | |
AK182824.1 | - | 4 - 5 | 106 | ||
AK217513.1 | - | 3 - 4 | 274 | 106 | |
BC076587.1 | - | 8 - 9 | 1601 | 106 |