The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001098404.1 | NP_001091874.1 | 1 - 2 | 309 | 112 |
NM_010936.3 | NP_035066.1 | 1 - 2 | 309 | 112 | |
GenBank mRNA | AF031814.1 | - | 1 - 2 | 124 | 112 |
AI746915.1 | - | 1 - 2 | 309 | 112 | |
AK011108.1 | - | 1 - 2 | 278 | 112 | |
AK018630.1 | - | 1 - 2 | 309 | 112 |