Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dmp1
Entrez Gene ID
13406
Gene Name
dentin matrix protein 1
Gene Aliases
AV020965, Dmp, PP
Chromosome Location
Chr.5: 104197558 - 104214109 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_016779.2 NP_058059.2 1 - 2 92 91
GenBank mRNA AK132177.1 - 1 - 2 93 91
AK157973.1 - 1 - 2 93 91
U65020.1 - 1 - 2 98 91

View Full Product Details