The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_177239.2 | NP_796213.2 | 7 - 8 | 526 | 82 |
GenBank mRNA | AK081684.1 | - | 7 - 8 | 501 | 82 |
AK164103.1 | - | 7 - 8 | 472 | 82 | |
AK173300.1 | - | 4 - 5 | 268 | 82 | |
BC150946.1 | - | 7 - 8 | 522 | 82 | |
BC151172.1 | - | 7 - 8 | 522 | 82 |