Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tet3
Entrez Gene ID
194388
Gene Name
tet methylcytosine dioxygenase 3
Gene Aliases
B430006D22Rik, BC037432, D230004J03Rik
Chromosome Location
Chr.6: 83362373 - 83457313 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_183138.2 NP_898961.2 7 - 8 2888 63
GenBank mRNA AK084164.1 - 2 - 3 241 63
BC096437.1 - 3 - 4 402 63
HQ423151.1 - 7 - 8 3062 63
HQ423152.1 - 6 - 7 3005 63
JX946278.1 - 8 - 9 3594 63

View Full Product Details