The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_183180.2 | NP_899003.1 | 8 - 9 | 1001 | 65 |
GenBank mRNA | AK032765.1 | - | 7 - 8 | 912 | 65 |
BC052169.1 | - | 8 - 9 | 964 | 65 |