The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_010463.1 | NP_034593.1 | 1 - 2 | 607 | 161 |
GenBank mRNA | AF448482.1 | - | 1 - 2 | 607 | 161 |
BC120845.1 | - | 1 - 2 | 703 | 161 | |
BC120847.1 | - | 1 - 2 | 703 | 161 |