The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001278534.1 | NP_001265463.1 | 2 - 2 | 4254 | 96 |
GenBank mRNA | AK032151.1 | - | 1 - 1 | 4579 | 96 |
AK220436.1 | - | 2 - 2 | 4251 | 96 | |
BB079282.1 | - | 1 - 1 | 214 | 96 | |
BP765965.1 | - | 1 - 1 | 96 |