Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Dnmbp
- Entrez Gene ID
-
71972
- Gene Name
- dynamin binding protein
- Gene Aliases
-
2410003L07Rik, 2410003M15Rik, TUBA
- Chromosome Location
-
Chr.19: 43846815 - 43940318 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AY383729.1 |
- |
1 - 2 |
156 |
88 |
View Full Product Details