Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dnmbp
Entrez Gene ID
71972
Gene Name
dynamin binding protein
Gene Aliases
2410003L07Rik, 2410003M15Rik, TUBA
Chromosome Location
Chr.19: 43846815 - 43940318 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK010367.1 - 3 - 4 301 59
AK129261.1 - 8 - 9 1046 59
AY383729.1 - 9 - 10 3177 59
BC025944.1 - 2 - 3 238 59

View Full Product Details