The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_198035.1 | NP_932152.1 | 2 - 2 | 5415 | 123 |
GenBank mRNA | AK033019.1 | - | 1 - 1 | 543 | 123 |
AK078957.1 | - | 1 - 1 | 1256 | 123 | |
AK172936.1 | - | 1 - 1 | 5205 | 123 | |
BC034437.1 | - | 1 - 1 | 2442 | 123 | |
BC058661.1 | - | 2 - 2 | 5415 | 123 | |
BC060725.1 | - | 1 - 1 | 5024 | 123 | |
BC079569.1 | - | 2 - 2 | 5311 | 123 |