The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022326.3 | NP_071721.2 | 7 - 8 | 993 | 127 |
GenBank mRNA | AF456463.1 | - | 6 - 7 | 902 | 127 |
AK005428.1 | - | 7 - 8 | 993 | 127 | |
AK005550.1 | - | 7 - 8 | 988 | 127 | |
AY014777.1 | - | 7 - 8 | 958 | 127 |