Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Emc2
Entrez Gene ID
66736
Gene Name
ER membrane protein complex subunit 2
Gene Aliases
4921531G14Rik, AV060620, AW209495, Ttc35
Chromosome Location
Chr.15: 43477214 - 43527777 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_025736.2 NP_080012.1 1 - 2 94 74
GenBank mRNA AK014989.1 - 1 - 2 95 74
AK015968.1 - 1 - 2 60 74
AK171442.1 - 1 - 2 83 74
AK172887.1 - 1 - 2 39 74
BC004716.1 - 1 - 2 52 74

View Full Product Details