The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_026532.3 | NP_080808.1 | 1 - 2 | 72 | 110 |
GenBank mRNA | AK020512.1 | - | 1 - 2 | 71 | 110 |
AK152212.1 | - | 1 - 2 | 82 | 110 | |
BC003955.1 | - | 1 - 2 | 72 | 110 | |
BC083165.1 | - | 1 - 2 | 126 | 110 |