The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001290514.1 | NP_001277443.1 | 3 - 4 | 471 | 103 |
NM_027409.5 | NP_081685.1 | 3 - 4 | 471 | 103 | |
NR_110964.1 | 3 - 4 | 471 | 103 | ||
GenBank mRNA | AK007533.1 | - | 3 - 4 | 404 | 103 |
AK088275.1 | - | 3 - 4 | 408 | 103 | |
AK139795.1 | - | 3 - 4 | 415 | 103 | |
BC018329.1 | - | 3 - 4 | 372 | 103 | |
BC085100.1 | - | 3 - 4 | 379 | 103 | |
BC100442.1 | - | 3 - 4 | 359 | 103 |