The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_133894.2 | NP_598655.1 | 3 - 4 | 1030 | 170 |
GenBank mRNA | AK034801.1 | - | 3 - 4 | 1031 | 170 |
BC015272.1 | - | 3 - 4 | 1027 | 170 | |
BC057169.1 | - | 3 - 4 | 1016 | 170 | |
BC069923.1 | - | 3 - 4 | 1018 | 170 |