The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013717.2 | NP_038745.1 | 3 - 4 | 279 | 71 |
GenBank mRNA | AB030483.1 | - | 3 - 4 | 279 | 71 |
AK003038.1 | - | 3 - 4 | 274 | 71 | |
AK010355.1 | - | 3 - 4 | 308 | 71 | |
AK131701.1 | - | 3 - 4 | 270 | 71 |