The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012024.2 | NP_036154.1 | 3 - 4 | 924 | 74 |
GenBank mRNA | AK088366.1 | - | 3 - 4 | 896 | 74 |
AK132432.1 | - | 3 - 4 | 957 | 74 | |
AK220163.1 | - | 3 - 4 | 914 | 74 | |
BC085149.1 | - | 3 - 4 | 924 | 74 | |
BC094387.1 | - | 3 - 4 | 916 | 74 | |
U49728.1 | - | 1 - 2 | 201 | 74 |