Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Hira
Entrez Gene ID
15260
Gene Name
histone cell cycle regulation defective homolog A (S. cerevisiae)
Gene Aliases
AA138857, D16Ertd95e, Gm15797, N28177, OTTMUSG00000026264, Tuple1
Chromosome Location
Chr.16: 18876750 - 18970309 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_010435.2 NP_034565.2 3 - 4 943 59
GenBank mRNA BC052856.1 - 3 - 4 361 59
BC060181.1 - 3 - 4 627 59
X75295.1 - 4 - 5 458 59
X92590.1 - 3 - 4 430 59
X99712.1 - 3 - 4 270 59

View Full Product Details